Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CCL22 (MDC) (67 aa) Recombinant Protein, PeproTech®

Catalog No. 30036500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30036500UG 500 μg
30036100UG 100 μg
300-36-1MG 1 mg
300-36-20UG 20 μg
30036250UG 250 μg
300-36-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30036500UG Supplier Gibco™ Supplier No. 30036500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MDC is an 8.0 kDa protein containing 67 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00626
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6367
Molecular Weight (g/mol) 8 kDa
Name Human CCL22 (MDC) (67 aa)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1
Common Name MDC
Gene Symbol CCL22
Biological Activity Determined by its ability to chemoattract human T cells using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence YGANMEDSVC CRDYVRYRLP LRVVKHFYWT SDSCPRPGVV LLTFRDKEIC ADPRVPWVKM ILNKLSQ
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.