Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CCL23 (P55773, 22 a.a. - 120 a.a.) Partial Recombinant Protein

Catalog No. 89938395 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-395 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-395 Supplier Abnova™ Supplier No. P3653
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist at CC chemokine receptor 1. Two alternatively spliced variants encoding different active isoforms have been identified. [provided by RefSeq]

Sequence: RVTKDAETEFMMSKLPLENPVLLDRFHATSADCCISYTPRSIPCSLLESYFETNSECSKPGVIFLTKKGRRFCANPSDKQVQVCMRMLKLDTRIKTRKN

Specifications

Accession Number P55773
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 6368
Molecular Weight (g/mol) 11kDa
Name CCL23 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Immunogen RVTKDAETEFMMSKLPLENPVLLDRFHATSADCCISYTPRSIPCSLLESYFETNSECSKPGVIFLTKKGRRFCANPSDKQVQVCMRMLKLDTRIKTRKN
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CK-BETA-8/CKb8/Ckb-8/Ckb-8-1/MIP-3/MIP3/MPIF-1/SCYA23
Common Name CCL23
Gene Symbol CCL23
Biological Activity Determined by its ability to chemoattract human T cells using a concentration range of 2.0-40.0ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.