Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CCL25/TECK Protein
SDP

Catalog No. 9046TK025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-46T-K025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-46T-K025 Supplier R&D Systems™ Supplier No. 9046TK025
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR9. The ED50 for this effect is 30-180 ng/mL.

Specifications

Accession Number O15444
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 6370
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions; M.W. (theoretical): 14 kDa
Quantity 25 μg
Source E. coli-derived human CCL25/TECK protein Gln24-Leu150
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Common Name CCL25/TECK
Gene Symbol CCL25
Biological Activity Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR9. The ED50 for this effect is typically 30-180ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in 4 mM HCl.
Expression System E. coli-derived Gln24-Leu150
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CCL25/TECK
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.