Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL4/MIP-1 beta Quantikine ELISA Kit

Catalog No. DMB00
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
DMB00 1 Plate 1 x 96-well Plate
SMB00 6 Plates 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DMB00 Supplier R&D Systems™ Supplier No. DMB00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human CCL4/MIP-1 beta Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL4/MIP-1 beta Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL4/MIP-1 beta in cell culture supernates (10 ul), serum (150 ul), edta plasma (150 ul), heparin plasma (150 ul), citrate plasma (150 ul).

Specifications

Assay Range 15.60 to 1000 pg/mL (Cell Culture Supernates), 31.20 to 2000 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Assay Sensitivity 4 pg/mL (Cell Culture Supernates), 11 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human MIP-1 beta
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6351
Quantity 1 Plate
Sample Volume Cell Culture Supernates (10uL),Serum (150uL),EDTA Plasma (150uL),Heparin Plasma (150uL),Citrate Plasma (150uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1 b), Small-inducible cytokine A4, T-cell activation protein 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.