Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL5/RANTES Quantikine ELISA Kit
SDP

Catalog No. SRN00B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SRN00B 6 Plates 6 x 96-well Plates
DRN00B 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SRN00B Supplier R&D Systems™ Supplier No. SRN00B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human CCL5/RANTES Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL5/RANTES Quantikine ELISA Kit quantifies human CCL5/RANTES in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 6.6pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity Natural and Recombinant Human RANTES
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6352
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.