Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CCL7 (MCP-3) Recombinant Protein

Catalog No. PIRP8649
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8649 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8649 Supplier Invitrogen™ Supplier No. RP8649

Recombinant Protein

CCL7 (human) Recombinant Protein, RP-8649, from Invitrogen.

Monocyte Chemotactic Protein 3 (MCP-3), also called CCL7, is produced by macrophages and some tumor cell lines. MCP-3 signals through three different G protein-coupled receptors, CCR1, CCR2, and CCR3. CCL7 chemoattracts monocytes and can regulate macrophage function. Recombinant human MCP-3 is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 9 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human PBMCs, human neutrophils or THP-1 cells and is typically 10-100 ng/mL. The specific activity of this protein is 1x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80098
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 6354
Molecular Weight (g/mol) 9
Name Human MCP-3
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-MCP-3
Common Name Human MCP-3
Gene Symbol CCL7
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitroresearch only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.