Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CCNB2 (NP_004692, 1 a.a. - 398 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89938478 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-478 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-478 Supplier Abnova™ Supplier No. P5843
Add to Cart
Add to Cart

Used for SDS-PAGE

Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMGSHMALLRRPTVSSDLENIDTGVNSKVKSHVTIRRTVLEEIGNRVTTRAAQVAKKAQNTKVPVQPTKTTNVNKQLKPTASVKPVQMEKLAPKGPSPTPEDVSMKEENLCQAFSDALLCKIEDIDNEDWENPQLCSDYVKDIYQYLRQLEVLQSINPHFLDGRDINGRMRAILVDWLVQVHSKFRLLQETLYMCVGIMDRFLQVQPVSRKKLQLVGITALLLASKYEEMFSPNIEDFVYITDNAYTSSQIREMETLILKELKFELGRPLPLHFLRRASKAGEVDVEQHTLAKYLMELTLIDYDMVHYHPSKVAAAASCLSQKVLGQGKWNLKQQYYTGYTENEVLEVMQHMAKNVVKVNENLTKFIAIKNKYASSKLLKISMIPQLNSKAVKDLASPLIGRS

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 9133
Molecular Weight (g/mol) 47.9kDa
Name CCNB2 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed.
Quantity 50 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGSHMALLRRPTVSSDLENIDTGVNSKVKSHVTIRRTVLEEIGNRVTTRAAQVAKKAQNTKVPVQPTKTTNVNKQLKPTASVKPVQMEKLAPKGPSPTPEDVSMKEENLCQAFSDALLCKIEDIDNEDWENPQLCSDYVKDIYQYLRQLEVLQSINPHFLDGRDINGRMRAILVDWLVQVHSKFRLLQETLYMCVGIMDRFLQVQPVSRKKLQLVGITALLLASKYEEMFSPNIEDFVYITDNAYTSSQIREMETLILKELKFELGRPLPLHFLRRASKAGEVDVEQHTLAKYLMELTLIDYDMVHYHPSKVAAAASCLSQKVLGQGKWNLKQQYYTGYTENEVLEVMQHMAKNVVKVNENLTKFIAIKNKYASSKLLKISMIPQLNSKAVKDLASPLIGRS
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HsT17299
Common Name CCNB2
Gene Symbol CCNB2
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS - PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.