Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Ccpg1 Full-Length Orf (Adz15580.1) Recombinant Protein Without Tag.

Catalog No. 89032118 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-118 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-118 Supplier Abnova™ Supplier No. H00009236G01
Add to Cart
Add to Cart

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Good Folding

Immunogen Sequence: MSTFFLIVFSSQPSDDVSSSDETSNQPSPAFRRRRARKKTVSASESEDRLVAEQETEPSKELSKRQFSSGLNKCVILALVIAISMGFGHFYGTIQIQKRQQLVRKIHEDELNDMKDYLSQCQQEQESFIDYKSLKENLARCWTLTEAEKMSFETQKTNLATENQYLRVSLEKEEKALSSLQEELNKLREQIRILEDKGTSTEL

Specifications

Accession Number ADZ15580.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 9236
Name CCPG1 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MSTFFLIVFSSQPSDDVSSSDETSNQPSPAFRRRRARKKTVSASESEDRLVAEQETEPSKELSKRQFSSGLNKCVILALVIAISMGFGHFYGTIQIQKRQQLVRKIHEDELNDMKDYLSQCQQEQESFIDYKSLKENLARCWTLTEAEKMSFETQKTNLATENQYLRVSLEKEEKALSSLQEELNKLREQIRILEDKGTSTELVKENQKLKQHLEEEKQKKHSFLSQRETLLTEAKMLKRELERERLVTTALRGELQQLSGSQLHGKSDSPNVYTEKKEIAILRERLTELELKLTFEQQRSDLWERLYVEAKDQNGKQGTDGKKKGGRGSHRAKNKSKETFLGSVKETFDAMKNSTKEFVRHHKEKIKQAKEAVKENLKKFSDSVKSTFRHFKDTTKNIFDEKGNKRFGATKEAAEKPRTVFSDYLHPQYKAPTENHHNRGPTMQNDGRKEKPVHFKEFRKNTNSKKCSPGHDCRENSHSFREACSGVFDCAQQESMSLFNIVVNPIRMDEFRQIIQRYMLKELDTFCHWNELDQFINKFFLNGVFIHDQKLFTDFVNDVKDYLRNMKEYEVDNDGVFEKLDEYIYRHFFGHTFSPPYGPRSVYIKPCHYSSL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CPR8/KIAA1254
Common Name CCPG1
Gene Symbol CCPG1
Species Wheat Germ (in vitro)
Protein Tag None
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.