Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7139666
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1791RSP 25 μg
MAB1791R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1791RSP Supplier R&D Systems Supplier No. MAB1791RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CCR9
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 248621R
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells, Immunocytochemistry 8-25μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P51686
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Host Species Mouse
Immunogen Human CCR9 transfectants Met1-Leu369 Accession # P51686
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 10803
Test Specificity Detects human CCR9 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.