Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CCRL2 Full-length ORF (AAH25717, 49 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89938554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-554 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-554 Supplier Abnova™ Supplier No. H00009034P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a chemokine receptor like protein, which is predicted to be a seven transmembrane protein and most closely related to CCR1. Chemokines and their receptors mediated signal transduction are critical for the recruitment of effector immune cells to the site of inflammation. This gene is expressed at high levels in primary neutrophils and primary monocytes, and is further upregulated on neutrophil activation and during monocyte to macrophage differentiation. The function of this gene is unknown. This gene is mapped to the region where the chemokine receptor gene cluster is located. [provided by RefSeq]

Sequence: FVIGVLDNLLVVLILVKYKGLKRAENIYLLNLAVSNLCFLLTLPFWAHAGGDPMCKILIGLYFVGLYSETFFNCLLTVQRYLVFLHKGNFFSARRRVPCGIITSVLAWVTAILATLPEYVVYKPQMEDQKYKCAFSRTPFLPADETFWKHFLTLKMNISVLVLPLFIFTFLYVQMRKTLRFREQRYSLFKLVFAIMVVFLLMWAPYNIAFFLSTFKEHFSLSDCKSSYNLDKSVHITKLIATTHCCINPLLYAFLDGTFSKYLCRCFHLRSNTPLQPRGQSAQGTSREEPDHSTEV

Specifications

Accession Number AAH25717
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9034
Molecular Weight (g/mol) 58.3kDa
Name CCRL2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen FVIGVLDNLLVVLILVKYKGLKRAENIYLLNLAVSNLCFLLTLPFWAHAGGDPMCKILIGLYFVGLYSETFFNCLLTVQRYLVFLHKGNFFSARRRVPCGIITSVLAWVTAILATLPEYVVYKPQMEDQKYKCAFSRTPFLPADETFWKHFLTLKMNISVLVLPLFIFTFLYVQMRKTLRFREQRYSLFKLVFAIMVVFLLMWAPYNIAFFLSTFKEHFSLSDCKSSYNLDKSVHITKLIATTHCCINPLLYAFLDGTFSKYLCRCFHLRSNTPLQPRGQSAQGTSREEPDHSTEV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CKRX/CRAM-A/CRAM-B/FLJ55815/HCR/MGC116710
Common Name CCRL2
Gene Symbol CCRL2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.