Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CD100 (soluble) Recombinant Protein, PeproTech®

Catalog No. 310291MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-29-1MG 1 mg
31029100UG 100 μg
310-29-20UG 20 μg
31029250UG 250 μg
31029500UG 500 μg
310-29-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 310-29-1MG Supplier Gibco™ Supplier No. 310291MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble CD100 is a 78.9 kDa protein comprising the extracellular domain of CD100 (711 amino acids). SDS-PAGE analysis run under non-reducing conditions shows a mixture of disulfide linked dimer and monomer. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92854
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10507
Molecular Weight (g/mol) 78.9 kDa
Name Human CD100 (soluble)
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4 D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4 D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4 D; semaphorin H; semaphorin-4 D; Semaphorin-C-like 2; semaphorin-J; Semcl2
Common Name CD100 (SEMA4D)
Gene Symbol Sema4d
Biological Activity Measured by its ability to inhibit chemokine (hMCP-3) induced human monocyte migration.
Conjugate Unconjugated
Recombinant Recombinant
Sequence FAPIPRITWE HREVHLVQFH EPDIYNYSAL LLSEDKDTLY IGAREAVFAV NALNISEKQH EVYWKVSEDK KAKCAEKGKS KQTECLNYIR VLQPLSATSL YVCGTNAFQP ACDHLNLTSF KFLGKNEDGK GRCPFDPAHS YTSVMVDGEL YSGTSYNFLG SEPIISRNSS HSPLRTEYAI PWLNEPSFVF ADVIRKSPDS PDGEDDRVYF FFTEVSVEYE FVFRVLIPRI ARVCKGDQGG LRTLQKKWTS FLKARLICSR PDSGLVFNVL RDVFVLRSPG LKVPVFYALF TPQLNNVGLS AVCAYNLSTA EEVFSHGKYM QSTTVEQSHT KWVRYNGPVP KPRPGACIDS EARAANYTSS LNLPDKTLQF VKDHPLMDDS VTPIDNRPRL IKKDVNYTQI VVDRTQALDG TVYDVMFVST DRGALHKAIS LEHAVHIIEE TQLFQDFEPV QTLLLSSKKG NRFVYAGSNS GVVQAPLAFC GKHGTCEDCV LARDPYCAWS PPTATCVALH QTESPSRGLI QEMSGDASVC PDKSKGSYRQ HFFKHGGTAE LKCSQKSNLA RVFWKFQNGV LKAESPKYGL MGRKNLLIFN LSEGDSGVYQ CLSEERVKNK TVFQVVAKHV LEVKVVPKPV VAPTLSVVQT EGSRIATKVL VASTQGSSPP TPAVQATSSG AITLPPKPAP TGTSCEPKIV INTVPQLHSE KTMYLKSSDN R
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.