Learn More
Human CD14 PE-conjugated Antibody, R&D Systems™



Description
Specifications
Specifications
| Antigen | CD14 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 134620 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P08571 |
| Gene Alias | CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein |
| Gene Symbols | CD14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.