Learn More
Sino Biological Human CD164 MAb 25TESTS 12260-MM05-P-25
Supplier:  Sino Biological 12260MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD164 (rh CD164; Catalog#12260-H08H; Q04900-1; Met1-Asp162) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-9463
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.