Learn More
R&D Systems™ Human CD164 (NP_006007) VersaClone cDNA
SDP
Catalog No. RDC0934 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC0934 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC0934 Supplier R&D Systems™ Supplier No. RDC0934
Add to Cart
Add to Cart
This item is not returnable. View return policy
CD164 molecule, sialomucin
The Human CD164 (NP_006007) VersaClone cDNA clone contains the complete ORF for CD164, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD164 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans