Learn More
ACROBiosystems Human CD20 500ug
Supplier:  ACROBiosystems CD0H52D5500UG
Encompass_Preferred
Human CD20 Full Length Protein FlagHis Tag (CD0-H52D5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).
Catalog No. 50-253-5395
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.