Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD200 His-tag Recombinant Protein

Catalog No. PIRP87981
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87981 1 mg
PIRP87980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP87981 Supplier Invitrogen™ Supplier No. RP87981
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstituted with sterile deionized water to 0.25 mg/mL. Concentration is measured by UV.

CD200 is a type-1 membrane glycoprotein in the immunoglobulin superfamily, featuring two immunoglobulin domains. It is widely expressed on various cells, including B and T cells, keratinocytes, nerve cells, follicular dendritic cells, ovarian cells, and trophoblasts. CD200 interacts with CD200R on myeloid cells, inhibiting macrophage activation, reducing IL-6 production, and attenuating lymphocyte activation. It also prevents mast cell degranulation and decreases TNF-alpha and IL-13 secretion, playing a role in immune regulation and maternal tolerance. Homologues of CD200 are found in herpesviruses and poxviruses, indicating a conserved immune modulation function. CD200 has multiple alternatively spliced transcript variants, contributing to its functional diversity. Diseases linked to CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy, highlighting its importance in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41217
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 4345
Molecular Weight (g/mol) 40-45 kDa
Name Human CD200 His-tag
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
Common Name CD200
Gene Symbol Cd200
Biological Activity Immobilized CD200 Protein, Human, Recombinant (His Tag) (at 2 μg/mL (100 μL/well) can bind CD200R Protein, Human, Recombinant (His and hFc Tag), the EC50 is 5-28 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Human CD200, amino acids Met1-Gly232 (Accession # NP_005935.4) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized CD200 Protein, Human, Recombinant (His Tag) at 2 μg/mL (100 μL/well) can bind CD200R Protein, Human, Recombinant (His and hFc Tag), the EC50 is 5-28 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >98% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.