Learn More
Invitrogen™ Human CD21/CR2/EBV receptor ELISA Kit
Description
Human CD21/CR2/EBV receptor quantitates human CD21/CR2/EBV receptor in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD21/CR2/EBV receptor.
Specifications
Specifications
| Accession Number | C3DR; CD21; CD35; cell surface receptor for C3d; complement C3d receptor; complement C3d receptor 2; complement component (3 b/4 b) receptor 1 (Knops blood group); complement component (3 d/Epstein Barr virus) receptor 2; complement component 3 d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; Complement receptor type 2; CR; Cr1; Cr-1; Cr2; Cr-2; CVID7; EBV receptor; Epstein-Barr virus receptor; expressed in B lymphocytes; SLEB9 |
| Assay Range | 17 to 4,000 pg/mL |
| Assay Sensitivity | 17 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 1380 |
| Gene Symbol | CR2 |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.