Learn More
Proteintech Group Inc Human CD23,FCER2 ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE0010796T
Encompass_Preferred
CD23, also known as low affinity immunoglobulin epsilon Fc receptor, is a transmembrane glycoprotein present on a subpopulation of B lymphocytes in germinal centres, EBV-transformed B-lymphoblastoid cell lines, follicular dendritic cells, and a subpopulation of peripheral blood cells. CD23 has essential roles in the regulation of IgE production and in the differentiation of B-cells. CD23 can be cleaved into soluble fragment (soluble CD23, sCD23) of various sizes displaying pleiotropic biological activities. Increased plasma levels of sCD23 have been reported in allergic disorders, acute viral diseases, lepromatous leprosy, B-CLL, and inflammatory diseases like RA, SLE and sarcoidosis.
Catalog No. 50-197-4067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.