Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CD244 Full-length ORF (NP_057466.1) Recombinant Protein without tag

Catalog No. 89938644 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-644 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-644 Supplier Abnova™ Supplier No. H00051744G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

Natural killer (NK) cells express both activating and inhibitory cell surface receptors. Inhibitory signaling receptors all possess cytoplasmic immunoreceptor tyrosine-based inhibitory motifs, or ITIMs, whereas the activating receptors lack ITIMs and associate with DAP12 (TYROBP; MIM 604142), which contains an immunoreceptor tyrosine-based activation motif, or ITAM. Killer cell immunoglobulin (Ig)-like receptors, or KIRs (see KIR2DL1; MIM 604936), and other NK cell receptors interact with major histocompatibility complex (MHC) molecules (see MIM 142800). Members of the CD2 (MIM 186990) family adhere to each other instead. The cell surface glycoprotein 2B4 is related to CD2 and is implicated in the regulation of NK- and T-cell function (Boles et al., 1999 [PubMed 10458320]).[supplied by OMIM]

Sequence: MLGQVVTLILLLLLKVYQGKGCQGSADHVVSISGVPLQLQPNSIQTKVDSIAWKKLLPSQNGFHHILKWENGSLPSNTSNDRFSFIVKNLSLLIKAAQQQDSGLYCLEVTSISGKVQTATFQVFVFDKVEKPRLQGQGKILDRGRCQVALSCLVSRDGNVSYAWYRGSKLIQTAGNLTYLDEEVDINGTHTYTCNVSNPVSWESHTLNLTQDCQNAHQEFRFWPFLVIIVILSALFLGTLACFCVWRRKRKEKQSETSPKEFLTIYEDVKDLKTRRNHEQEQTFPGGGSTIYSMIQSQSSAPTSQEPAYTLYSLIQPSRKSGSRKRNHSPSFNSTIYEVIGKSQPKAQNPARLSRKELENFDVYS

Specifications

Accession Number NP_057466.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 51744
Molecular Weight (g/mol) 41.1kDa
Name CD244 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MLGQVVTLILLLLLKVYQGKGCQGSADHVVSISGVPLQLQPNSIQTKVDSIAWKKLLPSQNGFHHILKWENGSLPSNTSNDRFSFIVKNLSLLIKAAQQQDSGLYCLEVTSISGKVQTATFQVFVFDKVEKPRLQGQGKILDRGRCQVALSCLVSRDGNVSYAWYRGSKLIQTAGNLTYLDEEVDINGTHTYTCNVSNPVSWESHTLNLTQDCQNAHQEFRFWPFLVIIVILSALFLGTLACFCVWRRKRKEKQSETSPKEFLTIYEDVKDLKTRRNHEQEQTFPGGGSTIYSMIQSQSSAPTSQEPAYTLYSLIQPSRKSGSRKRNHSPSFNSTIYEVIGKSQPKAQNPARLSRKELENFDVYS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 2B4/NAIL/NKR2B4/Nmrk/SLAMF4
Common Name CD244
Gene Symbol CD244
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.