Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD25/IL-2R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB623100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB623100 100 μg
MAB623SP 25 μg
MAB623500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB623100 Supplier R&D Systems Supplier No. MAB623100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CD25/IL-2R alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 24204
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01589
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha Glu22-Cys213 Accession # P01589
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Test Specificity Detects human CD25/IL-2 R alpha in ELISAs and Western blots. In Western blots, detects recombinant rat CD25/IL-2 R alpha but no cross-reactivity with recombinant mouse CD25/IL-2 R alpha . In sandwich immunoassays, this antibody does not cross-react or interfere with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-2 R beta , rhIL-3 R alpha , rhIL-4 R, rhIL-5 R alpha , rhIL-5 R beta , rhIL-6 R, or rhgp130.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.