Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CD27 Ligand (soluble) Recombinant Protein, PeproTech®

Catalog No. 3103010UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
310-30-10UG 10 μg
31030100UG 100 μg
310-30-1MG 1 mg
31030250UG 250 μg
31030500UG 500 μg
310-30-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 310-30-10UG Supplier Gibco™ Supplier No. 3103010UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's Recombinant Human sCD27L contains the extracellular domain plus an N-terminal His-Tag. The calculated molecular weight of Recombinant Human sCD27L is 18.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P32970
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 970
Molecular Weight (g/mol) 18.8 kDa
Name Human CD27 Ligand (soluble)
Quantity 10 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8 A; tumor necrosis factor ligand superfamily member 7
Common Name CD70
Gene Symbol Cd70
Biological Activity Determined by its ability to stimulate human IL-8 production by human PBMC using a concentration range of 10.0-25.0 ng/ml Note: Results may vary with PBMC donors.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence HHHHHHHHPS PGGSGGQRFA QAQQQLPLES LGWDVAELQL NHTGPQQDPR LYWQGGPALG RSFLHGPELD KGQLRIHRDG IYMVHIQVTL AICSSTTASR HHPTTLAVGI CSPASRSISL LRLSFHQGCT IASQRLTPLA RGDTLCTNLT GTLLPSRNTD ETFFGVQWVR P
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.