Learn More
Invitrogen™ Human CD27 Ligand/TNFSF7 ELISA Kit
Description
Human CD27 Ligand/TNFSF7 quantitates human CD27 Ligand/TNFSF7 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD27 Ligand/TNFSF7.
Specifications
Specifications
| Accession Number | CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8 A; tumor necrosis factor ligand superfamily member 7 |
| Assay Range | 0.3 to 70 ng/mL |
| Assay Sensitivity | 0.3 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 970 |
| Gene Symbol | Cd70 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.