Learn More
Sino Biological Human CD274 PAb 100UG 10084-RP02-B-100
Supplier:  Sino Biological 10084RP02B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B7-H1 / CD274 / PD-L1 (rh B7-H1; Catalog#10084-H08H; NP_054862.1; Met 1-Thr 239). B7-H1 specific IgG was purified by Human B7-H1 affinity chromatography and then biotinylated.
Catalog No. 50-240-9086
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.