Learn More
Sino Biological Human CD276 MAb 25TESTS 11188-MM06-A-25
Supplier:  Sino Biological 11188MM06A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human B7-H3 / CD276 (rh B7-H3 / CD276; Catalog#11188-H08H; Q5ZPR3-1; Met 1-Thr 461) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.