Learn More
Abcam Human CD3 Peptide, Synthetic, >90% Purity, 1 mg
Description
- Target Protein: CD3 epsilon
- Type: Synthetic blocking peptide
- Purity: >90%
- Tested Applications: BL
- Form: Liquid
- Species Reactivity: Human
- Storage Buffer: HEPES, 0.87% Sodium chloride, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Human |
| Target Molecule | CD3 epsilon (Human) |
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Buffer | HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.