Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CD30 Ligand (soluble) Recombinant Protein, PeproTech®

Catalog No. 4504210UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-42-10UG 10 μg
45042100UG 100 μg
450-42-1MG 1 mg
45042500UG 500 μg
450-42-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 450-42-10UG Supplier Gibco™ Supplier No. 4504210UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble CD30L (sCD30L) is a 188 amino acid polypeptide corresponding to the extracellular domain, and contains an 8 residue N-terminal His-Tag. The calculated molecular weight of Recombinant Human sCD30L is 21.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The ligand for CD30 is CD30L (CD153). The binding of CD30 to CD30L mediates pleiotropic effects including cell proliferation, activation, differentiation, and apoptotic cell death. CD30 has a critical role in the pathophysiology of Hodgkin's disease and other CD30+ lymphomas. CD30 acts as a costimulatory molecule in thymic negative selection. In addition to its expression on Hodgkin's and Reed-Sternberg cells, CD30 is also found in some non-Hodgkin's lymphomas (including Burkitt's lymphomas), virus-infected T and B cells, and on normal T and B cells after activation. In T cells, CD30 expression is present on a subset of T cells that produce Th2-type cytokines and on CD4+/CD8+ thymocytes that coexpress CD45RO and the IL4 receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P32971
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 944
Molecular Weight (g/mol) 21.3 kDa
Name Human CD30 Ligand (soluble)
Quantity 10 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3 A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8
Common Name CD153
Gene Symbol Tnfsf8
Biological Activity Determined by its ability to stimulate human IL-8 production by human PBMC using a concentration range of 10.0-25.0 ng/ml Note: Results may vary with PBMC donors.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence HHHHHHHHPS PGGSGGQRTD SIPNSPDNVP LKGGNCSEDL LCILKRAPFK KSWAYLQVAK HLNKTKLSWN KDGILHGVRY QDGNLVIQFP GLYFIICQLQ FLVQCPNNSV DLKLELLINK HIKKQALVTV CESGMQTKHV YQNLSQFLLD YLQVNTTISV NVDTFQYIDT STFPLENVLS IFLYSNSD
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.