Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD30 Ligand/TNFSF8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1028100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1028100 100 μg
MAB1028SP 25 μg
MAB1028500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1028100 Supplier R&D Systems Supplier No. MAB1028100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD30 Ligand/TNFSF8 Monoclonal specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen CD30 Ligand/TNFSF8
Applications Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 116614
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-4 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P32971
Gene Alias CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Gene Symbols TNFSF8
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8 Gln63-Asp234 Accession # P32971
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944
Test Specificity Detects human CD30 Ligand/TNFSF8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CD27 Ligand, recombinant mouse CD30 Ligand, or rhCD40 Ligand is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.