Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD300b/LMIR5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2879SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2879SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2879SP Supplier R&D Systems Supplier No. AF2879SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD300b/LMIR5/CD300LB Polyclonal specifically detects CD300b/LMIR5/CD300LB in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD300b/LMIR5/CD300LB
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_777552
Gene Alias CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CD300LB, CLM7, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Gm253, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, Triggering receptor expressed on myeloid cells 5
Gene Symbols CD300LB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD300b/LMIR5 Ile55-His187 Accession # NP_777552
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124599
Test Specificity Detects human CD300b/LMIR5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human LMIR2 and recombinant mouse LMIR5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.