Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD31/PECAM-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF806
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF806 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF806 Supplier R&D Systems Supplier No. AF806
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 25 publications

CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.

Specifications

Antigen CD31/PECAM-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated 5-15 ug/mL
Gene Alias adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Gene Symbols PECAM1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD31 Extracellular domain
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5175
Test Specificity Detects human CD31 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse CD31 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.