Learn More
Sino Biological Human CD3D/CD3E MAb 25TESTS CT026-R301-C-25
Supplier:  Sino Biological CT026R301C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D & CD3E (rh CD3D & CD3E; Catalog#CT026-H0323H; P04234-1 & P07766-1; Met1-Ala105 & Met1-Asp126) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-7883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.