Learn More
Invitrogen™ Human CD4 ELISA Kit
ELISA
$666.00 - $5890.00
Specifications
Accession Number | CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 receptor; CD4, allele 1; cd4a; CD4mut; cell surface glycoprotein CD4; fCD4; L3T4; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); W3/25; W3/25 antigen |
---|---|
Assay Range | 0.614 to 60 ng/mL |
Assay Sensitivity | 0.61 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH90RBX5
![]() |
Invitrogen™
EH90RBX5 |
5 x 96 Tests |
Each for $3,280.00
|
|
|||||
EH90RB
![]() |
Invitrogen™
EH90RB |
96 Tests |
Each for $666.00
|
|
|||||
EH90RBX10
![]() |
Invitrogen™
EH90RBX10 |
10 x 96 Tests |
Each for $5,890.00
|
|
|||||
Description
Human CD4 quantitates human CD4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD4.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 receptor; CD4, allele 1; cd4a; CD4mut; cell surface glycoprotein CD4; fCD4; L3T4; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); W3/25; W3/25 antigen | |
0.61 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CD4 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
CD4 | |
4 hrs 45 mins |
0.614 to 60 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
920 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.