Learn More
ACROBiosystems Human CD40 25tests
Supplier:  ACROBiosystems CD0HA25625TESTS
Encompass_Preferred
Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag (CD0-HA256) is produced via site-specific conjugation of AF647 to Human CD40 Protein Fc Tag under optimal conditions with a proprietary technology. Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).
Catalog No. 50-253-5393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.