Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD40 Ligand (TNFSF5) Recombinant Protein

Catalog No. PIRP8640
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8640 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8640 Supplier Invitrogen™ Supplier No. RP8640

Recombinant Protein

CD40L (human) Recombinant Protein, RP-8640, from Invitrogen.

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2. Recombinant human CD40 Ligand is a non-glycosylated protein, containing 149 amino acids, with a molecular weight of 16.3 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent production of IL-8 by human PBMCs and is typically 5-10 ng/mL or proliferation of human PBMCs and is typically 1 ug/mL. The specific activity of this protein is 1.3x10^5 units/mg or 1x10^6 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29965
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 10mM sodium phosphate with no preservative; pH 7.5
Gene ID (Entrez) 959
Molecular Weight (g/mol) 16.3
Name Human CD154 (CD40 Ligand)
pH Range 7.5
Quantity 50 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-CD-40-L
Common Name Human CD154 (CD40 Ligand)
Gene Symbol CD40LG
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitroresearch only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.