Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CD40 Ligand (TNFSF5) (soluble), Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF31002500
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF31002500 500 μg
AF31002100 100 μg
AF3100210UG 10 μg
AF310021MG 1 mg
AF3100250UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. AF31002500 Supplier Gibco™ Supplier No. AF31002500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble CD40 ligand is a 16.3 kDa protein containing 149 amino acid residues comprising the receptor binding TNF-like domain of CD40L. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29965
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 959
Molecular Weight (g/mol) 16.3 kDa
Name Human CD40 Ligand (TNFSF5) (soluble), Animal-Free
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Common Name CD154 (CD40 Ligand)
Gene Symbol CD40LG
Biological Activity Determined by the dose-dependent stimulation of IL-8 production by human PBMC. The expected ED50 for this effect is 5-10 ng/ml Note: Results may vary with PBMC donors.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.