Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CD40 Quantikine ELISA Kit

Catalog No. DCCD40
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DCCD40 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DCCD40 Supplier R&D Systems™ Supplier No. DCCD40
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sufficient for at least one 96-well plates

The Human CD40 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD40/TNFRSF5 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), saliva (100 ul), urine (20 ul), human milk (50 ul).

Specifications

Assay Range 19.50 to 1250 pg/mL
Assay Sensitivity 5.56 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human CD40
For Use With (Application) ELISA
Gene ID (Entrez) 958
Gene Symbol CD40
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Saliva (100uL),Urine (20uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Target CD40/TNFRSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.