Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD42b/GPIb alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4067
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4067 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4067 Supplier R&D Systems Supplier No. MAB4067
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD42b/GPIb alpha
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 486805
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P07359
Gene Alias Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Gene Symbols GP1BA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha His17-Leu505 Accession # P07359
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2811
Test Specificity Detects human CD42b/GPIb alpha in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.