Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD44v3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BBA11
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BBA11 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BBA11 Supplier R&D Systems Supplier No. BBA11
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CD44
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 3G5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Gene Symbols CD44
Host Species Mouse
Immunogen Recombinant human CD44v3-10
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Test Specificity Detects human CD44v3 in Western blots. The specificity of monoclonal antibody clone 3G5 was determined by FACS analysis on a panel of CD44 transfected COS cells, and was deduced to be specific for CD44 protein isoforms containing human variant exon 3 (9).
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.