Learn More
Sino Biological Human CD45RA MAb 25TESTS 68036-XM01-F-25
Supplier:  Sino Biological 68036XM01F25
Encompass_Preferred
The monoclonal antibody was purified by affinity chromatography. Then it was conjugated with FITC under optimum conditions, and the unreacted FITC was removed.
Catalog No. 50-241-1459
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.