Learn More
Sino Biological Human CD59 MAb 25TESTS 12474-R029-A-25
Supplier:  Sino Biological 12474R029A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD59 (rh CD59; Catalog#12474-H08H; P13987-1; Met1-Glu101) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.