Learn More
Sino Biological Recombinant Cynomolgus CD8A & CD8B Heterodimer Protein (Flag & His Tag)
Supplier:  Sino Biological CT077C2508H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD8A (XP_005575419.1) (Met42-Asp223) was fused with a flag tag at the C-terminus, constructed the plasmid 1 ; A DNA sequence encoding the cynomolgus CD8B (XP_005575417.1) (Met1-Pro170) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-222-3867
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.