Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD79 (CD79a/CD79b Heterodimer) His-Flag Tag Recombinant Protein

Catalog No. PIRP88014
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88014 100 μg
PIRP88015 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88014 Supplier Invitrogen™ Supplier No. RP88014
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1588 μL) be added to the vial to prepare a stock solution of 0.63 mg/mL. Concentration is measured by UV.

CD79a and CD79B (CD79b Molecule) are Protein Coding genes. Diseases associated with CD79B include Agammaglobulinemia 6, Autosomal Recessive and Agammaglobulinemia, Non-Bruton Type. Among its related pathways are Disease and Antigen activates B Cell Receptor (BCR) leading to generation of second messengers. Gene Ontology (GO) annotations related to this gene include transmembrane signaling receptor activity. An important paralog of this CD79B gene is CD79A.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11912, P40259
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 973, 974
Molecular Weight (g/mol) 37.47 kDa
Name Human CD79 (CD79a/CD79b Heterodimer) His-Flag Tag
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Common Name CD79a/CD79b
Gene Symbol CD79A, CD79B
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag DYKDDDDK-tag, His-tag
Sequence Human CD79A, amino acids Met1-Arg143 (Accession # NP_001774.1) with a C-terminal Flag-tag and human CD78B, amino acids Met1-Asp159 (Accession # NP_000617.1) with a C-terminal His-tag were co-expressed and purified as a human CD79A & CD79B heterodimer
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE. ≥85% as determined by SEC-HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.