Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD97 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7116509
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB25291RSP 25 μg
MAB2529R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB25291RSP Supplier R&D Systems Supplier No. MAB25291RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CD97 Isoform 1
Applications Neutralization, ELISA
Classification Monoclonal
Clone 380905R
Conjugate Unconjugated
Dilution Neutralization 0.25-1.25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_001775.2
Gene Alias CD97, TM7LN1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD97 Gln21-Gln398 Accession # NP_001775.2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 976
Test Specificity Detects human CD97 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.