Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CDC2 (NM_001786, 1 a.a. - 297 a.a.) and CCNA2 (NM_001237, 1 a.a. - 432 a.a.) Recombinant Protein with GST-tag expressed in Sf9 cells

Catalog No. 89938848 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-848 Supplier Abnova™ Supplier No. P4639
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. In contrast to cyclin A1, which is present only in germ cells, this cyclin is expressed in all tissues tested. This cyclin binds and activates CDC2 or CDK2 kinases, and thus promotes both cell cycle G1/S and G2/M transitions. [provided by RefSeq]

Specifications

Accession Number NM_001786 (Gene ID : 983);NM_001237 (Gene ID : 890)
Concentration 0.126 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 983/890
Molecular Weight (g/mol) CDC2: 63.882kDa, CCNA2
Name CDC2/CCNA2 (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST affinity chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias CDC28A/CDK1/DKFZp686L20222/MGC111195
Common Name CDC2
Gene Symbol CDC2
Biological Activity 8pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.