Learn More
Sino Biological Human CDH17 Protein 100UG 11360-H49H-B-100
Supplier:  Sino Biological 11360H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CDH17 (Q12864-1)(Met1-Met787) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-9487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.