Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CDH8 Full-length ORF (NP_001787.2, 1 a.a. - 799 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-001 25 ug
89-939-000 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-939-001 Supplier Abnova™ Supplier No. H00001006P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed in brain and is putatively involved in synaptic adhesion, axon outgrowth and guidance. [provided by RefSeq]

Sequence: MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKET

Specifications

Accession Number NP_001787.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1006
Molecular Weight (g/mol) 114.7kDa
Name CDH8 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKET
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Nbla04261
Common Name CDH8
Gene Symbol CDH8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.