Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CDK3 (NP_001249.1, 1 a.a. - 305 a.a.) and CCNE1 (NP_001229.1, 1 a.a. - 410 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells

Catalog No. 89939021 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-021 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-939-021 Supplier Abnova™ Supplier No. P5513
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK2, whose activity is required for cell cycle G1/S transition. This protein accumulates at the G1-S phase boundary and is degraded as cells progress through S phase. Overexpression of this gene has been observed in many tumors, which results in chromosome instability, and thus may contribute to tumorigenesis. This protein was found to associate with, and be involved in, the phosphorylation of NPAT protein (nuclear protein mapped to the ATM locus), which participates in cell-cycle regulated histone gene expression and plays a critical role in promoting cell-cycle progression in the absence of pRB. Two alternatively spliced transcript variants of this gene, which encode distinct isoforms, have been described. Two additional splice variants were reported but detailed nucleotide sequence information is not yet available. [provided by RefSeq]

Specifications

Accession Number NP_001249.1 (Gene ID : 1018);NP_001229.1 (Gene ID : 898)
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 1018/898
Molecular Weight (g/mol) 62kDa
Name CDK3/CCNE1 (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CCNE
Common Name CDK3
Gene Symbol CDK3
Biological Activity The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Modified Histone H1. ATP: 100 uM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 66% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.