Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CDKL3 Full-length ORF (AAH41799.1, 1 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939076 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-076 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-939-076 Supplier Abnova™ Supplier No. H00051265P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MEMYETLGKVGEGSYGTVMKCKHKNTGQIVAIKIFYERPEQSVNKIAMREIKFLKQFHHENLVNLIEVFRQKKKIHLVFEFIDHTVLDELQHYCHGLESKRLRKYLFQILRAIDYLHSNNIIHRDIKPENILVSQSGITKLCDFGFARTLAAPGDIYTDYVATRWYRAPELVLKDTSYGKPVDIWALGCMIIEMATGNPYLPSSSDLDLLHKIVLKVGNLSPHLQNIFSKSPIFAGVVLPQVQHPKNARKKYPKLNGLLADIVHACLQIDPADRISSSDLLHHEYFTRDGFIEKFMPELKAKLLQEAKVNSLIKPKESSKENELRKDERKTVYTNTLLSSSVLGKEIEKEKKPKEIKVRVIKVKGGRGDISEPKKKEYEGGLGQQDANENVHPMSPDTKLVTIEPPNPINPSTNCNGLKENPHCGGSVTMPPINLTNSNLMAANLSSNLFHPSVRLTERAKKRRTSSQSIGQVMPNSRQEDPGPIQSQMEKGIFNERTGHSDQMANENKRKLNFSRSDRKEFHFPELPVTIQSKDTKGMEVKQIKMLKRESKKTESSKIPTLLNVDQNQEKQEGGDGHCEGKNLKRNRFFFW

Specifications

Accession Number AAH41799.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51265
Molecular Weight (g/mol) 93.9kDa
Name CDKL3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MEMYETLGKVGEGSYGTVMKCKHKNTGQIVAIKIFYERPEQSVNKIAMREIKFLKQFHHENLVNLIEVFRQKKKIHLVFEFIDHTVLDELQHYCHGLESKRLRKYLFQILRAIDYLHSNNIIHRDIKPENILVSQSGITKLCDFGFARTLAAPGDIYTDYVATRWYRAPELVLKDTSYGKPVDIWALGCMIIEMATGNPYLPSSSDLDLLHKIVLKVGNLSPHLQNIFSKSPIFAGVVLPQVQHPKNARKKYPKLNGLLADIVHACLQIDPADRISSSDLLHHEYFTRDGFIEKFMPELKAKLLQEAKVNSLIKPKESSKENELRKDERKTVYTNTLLSSSVLGKEIEKEKKPKEIKVRVIKVKGGRGDISEPKKKEYEGGLGQQDANENVHPMSPDTKLVTIEPPNPINPSTNCNGLKENPHCGGSVTMPPINLTNSNLMAANLSSNLFHPSVRLTERAKKRRTSSQSIGQVMPNSRQEDPGPIQSQMEKGIFNERTGHSDQMANENKRKLNFSRSDRKEFHFPELPVTIQSKDTKGMEVKQIKMLKRESKKTESSKIPTLLNVDQNQEKQEGGDGHCEGKNLKRNRFFFW
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NKIAMRE
Common Name CDKL3
Gene Symbol CDKL3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.