Learn More
ACROBiosystems Human CEACAM-5 1mg
Supplier:  ACROBiosystems CE5H52H81MG
Encompass_Preferred
Human CEACAM-5 (499-685) Protein His Tag (CE5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 499 - Ala 685 (Accession # P06731-1).
Catalog No. 50-253-5304
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.