Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CEACAM-6/CD66c (NP_002474) VersaClone cDNA

Catalog No. RDC0955
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC0955 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC0955 Supplier R&D Systems™ Supplier No. RDC0955
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carcinoembryonic antigen-related cell adhesion molecule 6

The Human CEACAM-6/CD66c (NP_002474) VersaClone cDNA clone contains the complete ORF for CEACAM6/CD66c, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM6/CD66c insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

Specifications

Gene CEACAM6/CD66c
Accession Number NP_002474
Gene ID (Entrez) 4680
Gene Alias carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, CEACAM6, CEAL, NCA, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.