Learn More
Sino Biological Human CEACAM5 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 11077H27HB20
Encompass_Preferred
A DNA sequence encoding the human CEACAM5 (NP_004354.2) (Met1-Ala685) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.